Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NME1/NME2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589022
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589022 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589022 Supplier Invitrogen™ Supplier No. PA589022
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MANCERTFIA IKPDGVQRGL VGEIIKRFEQ KGFRLVGLKF MQASEDLLKE HYVDLKDRPF FAGLVKYMHS GPVVAMVWEG LNVVKTGRVM LGETNPADSK PGTIRGDFCI QVGRTMANLE RTFIAIKPDG VQRGLVGEII KRFEQKGFRL VAMKFLRASE EHLKQHYIDL KDRPFFPGLV KYMNSGPVVA MVWEGLNVVK TGRVMLGETN PADSKPGTIR GDFCIQVGRN IIHGSDSVKS AEKEISLWFK PEELVDYKSC AHDWVYE; Positive Samples: U-87MG, OVCAR-3, Mouse kidney, Rat kidney, Rat liver; Cellular Location: Cell projection, Cytoplasm, Nucleus, lamellipodium, ruffle.

TRUSTED_SUSTAINABILITY

Specifications

Antigen NME1/NME2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NME1
Gene Accession No. P15531, P15532, P19804, P22392, Q01768, Q05982
Gene Alias AL024257; AWD; C-myc purine-binding transcription factor PUF; c-myc transcription factor; epididymis secretory sperm binding protein Li 155an; expressed in non-metastatic cells 1 protein; expressed in non-metastatic cells 1 protein (NM23A) (nucleoside diphosphate kinase); expressed in non-metastatic cells 1, protein; expressed in non-metastatic cells 1, protein (NM23A) (nucleoside diphosphate kinase); expressed in non-metastatic cells 2; expressed in non-metastatic cells 2 protein; GAAD; Granzyme A-activated DNase; HEL-S-155an; histidine protein kinase NDKB; Metastasis inhibition factor nm23; NB; NBS; NDK A; NDK B; NDKA; NDKB; NDP kinase A; NDP kinase alpha; NDP kinase B; NDP kinase beta; NDPKA; NDPK-A; NDPKB; NDPK-B; NM23; nm23-2; NM23A; NM23B; NM23-H1; nm23-H2; NM23-M1; nm23-M2; NME/NM23 nucleoside diphosphate kinase 1; NME/NM23 nucleoside diphosphate kinase 2; NME1; NME2; non-metastatic cells 1, protein (NM23A) expressed in; non-metastatic cells 2, protein (NM23) expressed in; non-metastatic cells 2, protein (NM23B) expressed in; nucleoside diphosphate kinase A; Nucleoside diphosphate kinase B; nucleoside-diphosphate kinase 1; nucleoside-diphosphate kinase 2; nucleotide diphosphate kinase; nucleotide diphosphate kinase B; nucloside diphosphate kinase; P18; p18-12d; PUF; Tumor metastatic process-associated protein
Gene Symbols NME1, Nme2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18102, 18103, 191575, 4830, 4831, 83782
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.