Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NME1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143209
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143209 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5143209 Supplier Invitrogen™ Supplier No. PA5143209
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize both reported isoforms (NP_937818.1; NP_000260.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

NME1 was identified because of its reduced mRNA transcript levels in highly metastatic cells. Nucleoside diphosphate kinase (NDK) exists as a hexamer composed of 'A' (encoded by this gene) and 'B' (encoded by NME2) isoforms. Mutations in the gene have been identified in aggressive neuroblastomas. Two transcript variants encoding different isoforms have been found for this gene. Co-transcription of this gene and the neighboring downstream gene (NME2) generates naturally-occurring transcripts (NME1-NME2), which encodes a fusion protein comprised of sequence sharing identity with each individual gene product.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NME1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene NME1
Gene Accession No. P15531
Gene Alias AL024257; AWD; expressed in non-metastatic cells 1; expressed in non-metastatic cells 1 protein; expressed in non-metastatic cells 1 protein (NM23A) (nucleoside diphosphate kinase); expressed in non-metastatic cells 1, protein; expressed in non-metastatic cells 1, protein (NM23A) (nucleoside diphosphate kinase); GAAD; Granzyme A activated DNase (GAAD); Granzyme A-activated DNase; GZMA activated DNase; included; metastasis inhibition factor NM23; NB; NBR-A; NBR-B; NBS; NDK A; NDK A 1; NDK A 2; NDK NBR-A; NDK NBR-B; NDKA; NDP kinase A; NDP kinase A 1; NDP kinase A 2; NDP kinase beta; NDP kinase NBR-A; NDPKA; NDPK-A; NM23; NM23A; NM23-C1; NM23-H1; NM23H1B; nm23-M1; NME/NM23 nucleoside diphosphate kinase 1; Nme1; NME1-1; NME1-NME2 spliced read-through transcript; non-metastatic cells 1, protein (NM23A) expressed in; nucleoside diphosphate kinase A; nucleoside diphosphate kinase A 1; nucleoside diphosphate kinase A 2; nucleoside diphosphate kinase NBR-B; nucleoside-diphosphate kinase 1; nucleoside-diphosphate kinase NBR-A; nucleotide diphosphate kinase; nucloside diphosphate kinase; tumor metastatic process-associated protein
Gene Symbols NME1
Host Species Goat
Immunogen Peptide with sequence DYTSCAQNWIYE.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4830
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.