Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ NME2 (Human) Cell-Based ELISA Kit
Click to view available options
Quantity:
1 Kit
Description
NME2 (Human) Cell-Based ELISA Kit is an indirect enzyme-linked immunoassay for qualitative determination of NM23 expression in cultured cells.
Specifications
Specifications
Cross Reactivity | Human,Mouse,Rat |
Product Type | NME2 |
For Use With (Application) | Qualitative |
Gene Alias | MGC111212, NDPK-B, NDPKB, NM23-H2, NM23B, puf |
Gene ID (Entrez) | 4831/654364 |
Quantity | 1 Kit |
Regulatory Status | RUO |
Storage Requirements | Store at 4°C for six months. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction