Learn More
Invitrogen™ NME2 Monoclonal Antibody (4G7A8)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA1185
Description
MA1-185 detects NME2 from human samples. MA1-185 has been successfully tested in western blot and immunofluorescence applications. In western blot, MA1-185 detects NME2 (human) at the expected approximate molecular weight of 17kDa. The MA1-185 immunogen is full length NME2 expressed in E coli.
Nucleoside diphosphate kinase B is an enzyme that in humans is encoded by the NME2 gene. Nucleoside diphosphate kinase (NDK) exists as a hexamer composed of 'A' (encoded by NME1) and 'B' (encoded by this gene) isoforms. Multiple alternatively spliced transcript variants encoding the same isoform have been found for this gene. Co-transcription of this gene and the neighboring upstream gene (NME1) generates naturally-occurring transcripts (NME1-NME2) which encode a fusion protein consisting of sequence sharing identity with each individual gene product.
Specifications
NME2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide; pH 7.4 | |
P19804, P22392, Q01768 | |
NME2 | |
Purified, recombinant full length human NME2 (aa 1-152) expressed in E. Coli. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Western Blot, Immunocytochemistry | |
4G7A8 | |
Unconjugated | |
NME2 | |
2.7.13.3; c-myc purine-binding transcription factor PUF; c-myc transcription factor; epididymis secretory sperm binding protein Li 155an; expressed in non-metastatic cells 2; expressed in non-metastatic cells 2 protein; HEL-S-155an; histidine protein kinase NDKB; NDK B; NDKB; NDP kinase alpha; NDP kinase B; NDPKB; NDPK-B; nm23-2; NM23B; nm23-H2; nm23-M2; NME/NM23 nucleoside diphosphate kinase 2; Nme2; non-metastatic cells 2, protein (NM23) expressed in; non-metastatic cells 2, protein (NM23B) expressed in; nucleoside diphosphate kinase B; nucleoside-diphosphate kinase 2; nucleotide diphosphate kinase B; P18; p18-12d; PUF | |
Mouse | |
Protein A | |
RUO | |
18103, 4831, 83782 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.