Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NME2 Recombinant Rabbit Monoclonal Antibody (7L3A2)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA535687
Description
Immunogen sequence: QHYIDLKDRP FFPGLVKYMN SGPVVAMVWE GLNVVKTGRV MLGETNPADS KPGTIRGDFC IQVGRNIIHG SDSVKSAEKE ISLWFKPEEL VDYKSCAHDW VYE.
Nucleoside diphosphate kinase B is an enzyme that in humans is encoded by the NME2 gene. Nucleoside diphosphate kinase (NDK) exists as a hexamer composed of 'A' (encoded by NME1) and 'B' (encoded by this gene) isoforms. Multiple alternatively spliced transcript variants encoding the same isoform have been found for this gene. Co-transcription of this gene and the neighboring upstream gene (NME1) generates naturally-occurring transcripts (NME1-NME2) which encode a fusion protein consisting of sequence sharing identity with each individual gene product.
Specifications
NME2 | |
Recombinant Monoclonal | |
0.24 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P19804, Q01768 | |
NME2 | |
A synthetic peptide corresponding to a sequence within amino acids 50-152 of human NME2/NM23B (P22392). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
7L3A2 | |
Unconjugated | |
NME2 | |
2.7.13.3; c-myc purine-binding transcription factor PUF; c-myc transcription factor; epididymis secretory sperm binding protein Li 155an; expressed in non-metastatic cells 2; expressed in non-metastatic cells 2 protein; HEL-S-155an; histidine protein kinase NDKB; NDK B; NDKB; NDP kinase alpha; NDP kinase B; NDPKB; NDPK-B; nm23-2; NM23B; nm23-H2; nm23-M2; NME/NM23 nucleoside diphosphate kinase 2; Nme2; non-metastatic cells 2, protein (NM23) expressed in; non-metastatic cells 2, protein (NM23B) expressed in; nucleoside diphosphate kinase B; nucleoside-diphosphate kinase 2; nucleotide diphosphate kinase B; P18; p18-12d; PUF | |
Rabbit | |
Affinity chromatography | |
RUO | |
18103, 83782 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction