Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NME2 Mouse anti-Human, Clone:4G7A8, Abnova™

Catalog No. 89333104 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-104 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-104 Supplier Abnova Corporation Supplier No. MAB16654
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human NME2.

Nucleoside diphosphate kinase (NDK) exists as a hexamer composed of ′A′ (encoded by NME1) and ′B′ (encoded by this gene) isoforms. Multiple alternatively spliced transcript variants encoding the same isoform have been found for this gene. Co-transcription of this gene and the neighboring upstream gene (NME1) generates naturally-occurring transcripts (NME1-NME2) which encode a fusion protein comprised of sequence sharing identity with each individual gene product. [provided by RefSeq]

Specifications

Antigen NME2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4G7A8
Conjugate Unconjugated
Description non-metastatic cells 2, protein (NM23B) expressed in
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry (1:200-1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene NME2
Gene Alias MGC111212, NDPK-B, NDPKB, NM23-H2, NM23B, puf
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 1-152 of human NME2 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 4831
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.