Learn More
NME2 Mouse anti-Human, Clone: OTI2F9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505574

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Nucleoside diphosphate kinase B is an enzyme that in humans is encoded by the NME2 gene. Nucleoside diphosphate kinase (NDK) exists as a hexamer composed of ′A′ (encoded by NME1) and ′B′ (encoded by this gene) isoforms. Multiple alternatively spliced transcript variants encoding the same isoform have been found for this gene. Co-transcription of this gene and the neighboring upstream gene (NME1) generates naturally-occurring transcripts (NME1-NME2) which encode a fusion protein consisting of sequence sharing identity with each individual gene product.Specifications
NME2 | |
Monoclonal | |
Unconjugated | |
NME2 | |
NDKB, NDPK-B, NDPKB, NM23-H2, NM23B, PUF | |
Mouse | |
Affinity Chromatography | |
RUO | |
4831 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2F9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P22392 | |
NME2 | |
Full length human recombit protein of human NME2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.