Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NME2 + NM23A Rabbit anti-Human, Non-human Primate, Polyclonal, FabGennix

Catalog No. 502207907
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-7907 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-7907 Supplier Fabgennix Inc Supplier No. NDKAB112AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NME2 + NM23A Polyclonal antibody specifically detects NME2 + NM23A in Human, Non-human Primate samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen NME2 + NM23A
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene Nme2
Gene Accession No. P15531, P22392
Gene Alias AL024257; AWD; C-myc purine-binding transcription factor PUF; c-myc transcription factor; epididymis secretory sperm binding protein Li 155an; expressed in non-metastatic cells 1 protein; expressed in non-metastatic cells 1 protein (NM23A) (nucleoside diphosphate kinase); expressed in non-metastatic cells 1, protein; expressed in non-metastatic cells 1, protein (NM23A) (nucleoside diphosphate kinase); expressed in non-metastatic cells 2; expressed in non-metastatic cells 2 protein; GAAD; Granzyme A-activated DNase; HEL-S-155an; histidine protein kinase NDKB; Metastasis inhibition factor nm23; NB; NBS; NDK A; NDK B; NDKA; NDKB; NDP kinase A; NDP kinase alpha; NDP kinase B; NDP kinase beta; NDPKA; NDPK-A; NDPKB; NDPK-B; NM23; nm23-2; NM23A; NM23B; NM23-H1; nm23-H2; NM23-M1; nm23-M2; NME/NM23 nucleoside diphosphate kinase 1; NME/NM23 nucleoside diphosphate kinase 2; NME1; NME2; non-metastatic cells 1, protein (NM23A) expressed in; non-metastatic cells 2, protein (NM23) expressed in; non-metastatic cells 2, protein (NM23B) expressed in; nucleoside diphosphate kinase A; Nucleoside diphosphate kinase B; nucleoside-diphosphate kinase 1; nucleoside-diphosphate kinase 2; nucleotide diphosphate kinase; nucleotide diphosphate kinase B; nucloside diphosphate kinase; P18; p18-12d; PUF; Tumor metastatic process-associated protein
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 100-150 on NM23A protein selective for both NDKA and NDKB.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100533953, 4830, 4831
Target Species Human, Non-human Primate
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.