Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NME4 Antibody (OTI1A5), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273005PEATT594
Description
NME4 Monoclonal specifically detects NME4 in Human samples. It is validated for Flow Cytometry.Specifications
NME4 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
NME4 | |
Full length recombinant protein of human NME4 (NP_005000) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2b |
Flow Cytometry | |
OTI1A5 | |
Flow Cytometry | |
EC 2.7.4.6, NDP kinase, mitochondrial, NDPK-D, NM23D, nm23-H4mitochondrial, non-metastatic cells 4, protein expressed in | |
Mouse | |
Affinity Purified | |
Core ESC Like Genes, Lipid and Metabolism, Stem Cell Markers | |
4833.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction