Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NME4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544586
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544586 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544586 Supplier Invitrogen™ Supplier No. PA544586
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VAMVWEGYNV VRASRAMIGH TDSAEAAPGT IRGDFSVHIS RNVIHASDSV Sequence homology: Cow: 86%; Dog: 100%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 93%; Zebrafish: 79%.

NME4 is a protein kinases, which are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The AGC kinase group consists of 63 kinases including the cyclic nucleotide-regulated protein kinase (PKA PKG) family, the diacylglycerol-activated/phospholipid-dependent protein kinase C (PKC) family, the related to PKA and PKC (RAC/Akt) protein kinase family, the kinases that phosphorylate G protein-coupled receptors family (ARK), and the kinases that phosphorylate ribosomal protein S6 family (RSK).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NME4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NME4
Gene Accession No. O00746
Gene Alias 2610027N22Rik; 2810024O08Rik; 5730493H09Rik; expressed in non-metastatic cells 4, protein; NDK; NDP kinase D; NDP kinase, mitochondrial; NDPKD; NDPK-D; NM23D; NM23H4; nm23-H4; Nm23M4; NM23-M4; NM23-Z4; NME/NM23 nucleoside diphosphate kinase 4; Nme4; non-metastatic cells 4, protein expressed in; nucleoside diphosphate kinase 4; nucleoside diphosphate kinase D; Nucleoside diphosphate kinase, mitochondrial; Unknown (protein for MGC:134183); zgc:56482
Gene Symbols NME4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human NME4.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4833
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.