Learn More
NME5 Monoclonal Antibody (4F5)
NME5 Monoclonal Antibody (4F5)
Supplier:  Thermo Scientific MA520379
Catalog No. PIMA520379
Only null left
Add to Cart
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NME5
Monoclonal
0.2 to 1.0 mg/mL
PBS with no preservative; pH 7.4
P56597
NME5
NME5 (NP_003542, 121 a.a. ~ 212 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
ELISA, Immunoprecipitation, Western Blot
4F5
Unconjugated
NME5
1700019D05Rik; expressed in non-metastatic cells 5; inhibitor of p53-induced apoptosis-beta; IPIA-beta; NDK-H 5; NDP kinase homolog 5; NM23H5; NM23-H5; nm23-M5; NME/NM23 family member 5; NME5; non-metastatic cells 5 protein expressed in; non-metastatic cells 5, protein expressed in (nucleoside-diphosphate kinase); nucleoside diphosphate kinase homolog 5; radial spoke 23 homolog; RSPH23; testis-specific nm23 homolog
Mouse
Affinity chromatography
RUO
8382
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.