Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NME5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140227
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140227 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140227 Supplier Invitrogen™ Supplier No. PA5140227
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LVAMILARHK AISYWLELLG PNNSLVAKET HPDSLRAIYG TDDLRNALHG SNDFAAAERE IR Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 90%.

Does not seem to have NDK kinase activity. Confers protection from cell death by Bax and alters the cellular levels of several antioxidant enzymes including Gpx5. May play a role in spermiogenesis by increasing the ability of late-stage spermatids to eliminate reactive oxygen species.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NME5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NME5
Gene Accession No. P56597
Gene Alias 1700019D05Rik; expressed in non-metastatic cells 5; inhibitor of p53-induced apoptosis-beta; IPIA-beta; NDK-H 5; NDP kinase homolog 5; NM23H5; NM23-H5; nm23-M5; NME/NM23 family member 5; NME5; non-metastatic cells 5 protein expressed in; non-metastatic cells 5, protein expressed in (nucleoside-diphosphate kinase); nucleoside diphosphate kinase homolog 5; radial spoke 23 homolog; RSPH23; testis-specific nm23 homolog
Gene Symbols NME5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NME5. Recombinant protein control fragment (Product #RP-108952).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8382
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.