Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NME5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$191.80 - $414.50
Specifications
Antigen | NME5 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEPA5145412
|
Invitrogen™
PA5145412 |
20 μL |
Each for $191.80
|
|
|||||
FEPA5145411
|
Invitrogen™
PA5145411 |
200 μL |
Each for $414.50
|
|
|||||
Description
For immunocytochemistry the cells were formalin fixed.
Does not seem to have NDK kinase activity. Confers protection from cell death by Bax and alters the cellular levels of several antioxidant enzymes including Gpx5. May play a role in spermiogenesis by increasing the ability of late-stage spermatids to eliminate reactive oxygen species.Specifications
NME5 | |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Rat | |
Q99MH5 | |
688903, 75533 | |
Recombinant Mouse NME5 protein, Leu16-Glu210 (Accession #Q99MH5). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
1700019D05Rik; expressed in non-metastatic cells 5; inhibitor of p53-induced apoptosis-beta; IPIA-beta; NDK-H 5; NDP kinase homolog 5; NM23H5; NM23-H5; nm23-M5; NME/NM23 family member 5; NME5; non-metastatic cells 5 protein expressed in; non-metastatic cells 5, protein expressed in (nucleoside-diphosphate kinase); nucleoside diphosphate kinase homolog 5; radial spoke 23 homolog; RSPH23; testis-specific nm23 homolog | |
NME5 | |
IgG | |
Protein A | |
NME5 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title