Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NME5 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | NME5 |
---|---|
Dilution | Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
NME5 Polyclonal antibody specifically detects NME5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
NME5 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Apoptosis | |
PBS, pH 7.2, 40% glycerol | |
8382 | |
IgG | |
Affinity purified |
Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Inhibitor of p53-induced apoptosis-beta, IPIA-beta, NDK-H 5, NDP kinase homolog 5, nm23-H5, NM23H5, non-metastatic cells 5, protein expressed in (nucleoside-diphosphate kinase), nucleoside diphosphate kinase homolog 5, radial spoke 23 homolog, RSPH23, Testis-specific nm23 homolog | |
This antibody was developed against Recombinant Protein corresponding to amino acids: LVAMILARHKAISYWLELLGPNNSLVAKETHPDSLRAIYGTDDLRNALHGSNDFAAAEREIR | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title