Learn More
NMI Mouse anti-Human, Clone: OTI2C1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809982

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
NMYC interactor (NMI) encodes a protein that interacts with NMYC and CMYC (two members of the oncogene Myc family), and other transcription factors containing a Zip, HLH, or HLH-Zip motif. The NMI protein also interacts with all STATs except STAT2 and augments STAT-mediated transcription in response to cytokines IL2 and IFN-gamma. The NMI mRNA has low expression levels in all human fetal and adult tissues tested except brain and has high expression in cancer cell line-myeloid leukemias.Specifications
NMI | |
Monoclonal | |
Unconjugated | |
NMI | |
N-myc and STAT interactor | |
Mouse | |
Affinity Chromatography | |
RUO | |
9111 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2C1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q13287 | |
NMI | |
Human recombit protein fragment corresponding to amino acids 1-223 of human NMI produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.