Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NMI Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552584
Description
Immunogen sequence: KINVTEIPDT LREDQMRDKL ELSFSKSRNG GGEVDRVDYD RQSGSAVITF VEIGVADKIL KKKEYPLYIN QTCHRVTVSP YTEIHLKKYQ IFSGTSKRTV LLTGMEGIQM DEEIVEDLIN IHFQRAKNGG GEVDVVKCSL GQPH Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 46%.
NMYC interactor (NMI) encodes a protein that interacts with NMYC and CMYC (two members of the oncogene Myc family), and other transcription factors containing a Zip, HLH, or HLH-Zip motif. The NMI protein also interacts with all STATs except STAT2 and augments STAT-mediated transcription in response to cytokines IL2 and IFN-gamma. The NMI mRNA has low expression levels in all human fetal and adult tissues tested except brain and has high expression in cancer cell line-myeloid leukemias.
Specifications
| NMI | |
| Polyclonal | |
| Unconjugated | |
| NMI | |
| N-mcy (and STAT) interactor; Nmi; N-myc (and STAT) interactor; N-myc and STAT interactor; N-myc interactor; N-myc-interactor | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9111 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13287 | |
| NMI | |
| Recombinant protein corresponding to Human NMI. Recombinant protein control fragment (Product #RP-89342). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction