Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NMNAT-1 Antibody (OTI1G4), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273017FR
Description
NMNAT-1 Monoclonal specifically detects NMNAT-1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
NMNAT-1 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
NMNAT1 | |
Full length recombinant protein of human NMNAT-1 (NP_073624) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI1G4 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
EC 2.7.7.1, EC 2.7.7.18, NaMN adenylyltransferase 1, nicotinamide mononucleotide adenylyltransferase 1, nicotinamide nucleotide adenylyltransferase, nicotinamide nucleotide adenylyltransferase 1, Nicotinate-nucleotide adenylyltransferase 1, NMN adenylyltransferase 1, NMNATPNAT1, pyridine nucleotide adenylyltransferase 1 | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
64802.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction