Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NMNAT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584418
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584418 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584418 Supplier Invitrogen™ Supplier No. PA584418
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LIPAYHRVIM AELATKNSKW VEVDTWESLQ KEWKETLKVL RHHQEKLEAS DCDHQQNSPT LERPGRKRKW TETQD Highest antigen sequence indentity to the following orthologs: Mouse - 72%, Rat - 75%.

The coenzyme NAD and its derivatives are involved in hundreds of metabolic redox reactions and are utilized in protein ADP-ribosylation, histone deacetylation, and in some Ca(2+) signaling pathways. NMNAT (EC 2.7.7.1) is a central enzyme in NAD biosynthesis, catalyzing the condensation of nicotinamide mononucleotide (NMN) or nicotinic acid mononucleotide (NaMN) with the AMP moiety of ATP to form NAD or NaAD.The coenzyme NAD and its derivatives are involved in hundreds of metabolic redox reactions and are utilized in protein ADP-ribosylation, histone deacetylation, and in some Ca(2+) signaling pathways. NMNAT (EC 2.7.7.1) is a central enzyme in NAD biosynthesis, catalyzing the condensation of nicotinamide mononucleotide (NMN) or nicotinic acid mononucleotide (NaMN) with the AMP moiety of ATP to form NAD or NaAD (Zhang et al., 2003 [PubMed 12574164]).[supplied by OMIM]. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMNAT1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NMNAT1
Gene Accession No. Q9HAN9
Gene Alias 2610529L11Rik; 5730441G13Rik; D4Cole1e; deamido-NAD(+) diphosphorylase; deamido-NAD(+) pyrophosphorylase; EC 2.7.7.1; EC 2.7.7.18; id:ibd5068; im:7144541; LCA9; NAD(+) diphosphorylase; NAD(+) pyrophosphorylase; naMN adenylyltransferase 1; nicotinamide mononucleotide adenylyl transferase; nicotinamide mononucleotide adenylyl transferase 1; nicotinamide mononucleotide adenylyltransferase 1; nicotinamide mononucleotide adenylyltransferase 1-like protein; nicotinamide nucleotide adenylyltransferase 1; nicotinamide/nicotinate mononucleotide adenylyltransferase 1; nicotinamide/nicotinic acid mononucleotide adenylyltransferase 1; nicotinamide-nucleotide adenylyltransferase 1; Nicotinate-nucleotide adenylyltransferase 1; NMN adenylyltransferase 1; NMN/NaMN adenylyltransferase 1; nmnat; NMNAT1; NMNATPNAT1; PNAT1; PNAT-1; pyridine nucleotide adenylyltransferase 1; zgc:110243
Gene Symbols NMNAT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NMNAT1. Recombinant protein control fragment (Product #RP-101965).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64802
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.