Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NMNAT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115662
Description
Antibody detects endogenous levels of total NMNAT2.
Catalyzes the formation of NAD(+) from nicotinamide mononucleotide (NMN) and ATP. Can also use the deamidated form; nicotinic acid mononucleotide (NaMN) as substrate but with a lower efficiency. Cannot use triazofurin monophosphate (TrMP) as substrate. Also catalyzes the reverse reaction, i.e. the pyrophosphorolytic cleavage of NAD(+). For the pyrophosphorolytic activity prefers NAD(+), NADH and NAAD as substrates and degrades nicotinic acid adenine dinucleotide phosphate (NHD) less effectively. Fails to cleave phosphorylated dinucleotides NADP(+), NADPH and NAADP(+).
Specifications
NMNAT2 | |
Polyclonal | |
Unconjugated | |
NMNAT2 | |
AI843915; Blad; C1orf15; D030041I09Rik; KIAA0479; naMN adenylyltransferase 1; naMN adenylyltransferase 2; Nicotinamide mononucleotide adenylyltransferase 2; nicotinamide nucleotide adenylyltransferase 2; nicotinamide/nicotinic acid mononucleotide adenylyltransferase 2; nicotinate-nucleotide adenylyltransferase 1; nicotinate-nucleotide adenylyltransferase 2; NMN adenylyltransferase 2; NMN/NaMN adenylyltransferase 2; Nmnat2; PNAT1; PNAT2; Protein bloated bladder; pyridine nucleotide adenylyltransferase 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
226518, 23057, 289095 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q0HA29, Q8BNJ3, Q9BZQ4 | |
NMNAT2 | |
A synthesized peptide derived from human NMNAT2(Accession Q9BZQ4), corresponding to amino acid residues K249-S299. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction