Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NMNAT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139759
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139759 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139759 Supplier Invitrogen™ Supplier No. PA5139759
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: TGRFIVIGGI VSPVHDSYGK QGLVSSRHRL IMCQLAVQNS DWIRVDPWEC YQDTWQTTCS VLEHHRDLMK RVTGCILSNV NTPSMTPVIG QPQNETPQPI YQNSNVATKP TAAKILGKVG ESLSRICCVR PPVERFTFVD ENANLGTVM Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 97%.

Catalyzes the formation of NAD(+) from nicotinamide mononucleotide (NMN) and ATP. Can also use the deamidated form; nicotinic acid mononucleotide (NaMN) as substrate but with a lower efficiency. Cannot use triazofurin monophosphate (TrMP) as substrate. Also catalyzes the reverse reaction, i.e. the pyrophosphorolytic cleavage of NAD(+). For the pyrophosphorolytic activity prefers NAD(+), NADH and NAAD as substrates and degrades nicotinic acid adenine dinucleotide phosphate (NHD) less effectively. Fails to cleave phosphorylated dinucleotides NADP(+), NADPH and NAADP(+).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMNAT2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NMNAT2
Gene Accession No. Q9BZQ4
Gene Alias AI843915; Blad; C1orf15; D030041I09Rik; KIAA0479; naMN adenylyltransferase 1; naMN adenylyltransferase 2; Nicotinamide mononucleotide adenylyltransferase 2; nicotinamide nucleotide adenylyltransferase 2; nicotinamide/nicotinic acid mononucleotide adenylyltransferase 2; nicotinate-nucleotide adenylyltransferase 1; nicotinate-nucleotide adenylyltransferase 2; NMN adenylyltransferase 2; NMN/NaMN adenylyltransferase 2; Nmnat2; PNAT1; PNAT2; Protein bloated bladder; pyridine nucleotide adenylyltransferase 2
Gene Symbols NMNAT2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NMNAT2. Recombinant protein control fragment (Product #RP-90847).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23057
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.