Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NMNAT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563436
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563436 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563436 Supplier Invitrogen™ Supplier No. PA563436
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VDPWESEQAQ WMETVKVLRH HHSKLLRSPP QMEGPDHGKA LFSTPAAVPE LKLLCGADVL KTFQTPNLWK DAHIQEIVEK FGLVCV Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 83%.

Catalyzes the formation of NAD(+) from nicotinamide mononucleotide (NMN) and ATP. Can also use the deamidated form; nicotinic acid mononucleotide (NaMN) as substrate with the same efficiency. Can use triazofurin monophosphate (TrMP) as substrate. Can also use GTP and ITP as nucleotide donors. Also catalyzes the reverse reaction, i.e. the pyrophosphorolytic cleavage of NAD(+). For the pyrophosphorolytic activity, can use NAD (+), NADH, NAAD, nicotinic acid adenine dinucleotide phosphate (NHD), nicotinamide guanine dinucleotide (NGD) as substrates. Fails to cleave phosphorylated dinucleotides NADP(+), NADPH and NAADP(+). Protects against axonal degeneration following injury.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMNAT3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NMNAT3
Gene Accession No. Q96T66
Gene Alias 4933408N02Rik; FKSG76; naMN adenylyltransferase 1; naMN adenylyltransferase 3; nicotinamide mononucleotide adenylyltransferase 3; nicotinamide nucleotide adenylyltransferase 3; Nicotinamide/nicotinic acid mononucleotide adenylyltransferase 3; nicotinamide-nucleotide adenylyltransferase 3; nicotinate-nucleotide adenylyltransferase 1; nicotinate-nucleotide adenylyltransferase 3; NMN adenylyltransferase 3; NMN/NaMN adenylyltransferase 3; NMNAT3; PNAT3; PNAT-3; pyridine nucleotide adenylyltransferase 3
Gene Symbols NMNAT3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NMNAT3. Recombinant protein control fragment (Product #RP-96415).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 349565
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.