Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMP (1-Methyl-2-pyrrolidinone), BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02003527
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Narrow Mouth Amber Glass Bottle

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
Synonym 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
PubChem CID 13387
ChEBI CHEBI:7307
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

Quantity 4 L
Flash Point 96.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.03kg
Formula Weight 99.13
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material NMP (1-Methyl-2-pyrrolidinone)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.