Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NMRAL1 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299301MFV500
Description
NMRAL1 Polyclonal antibody specifically detects NMRAL1 in Human samples. It is validated for Western BlotSpecifications
NMRAL1 | |
Polyclonal | |
Western Blot | |
HSCARGFLJ25918, NmrA-like family domain containing 1, nmrA-like family domain-containing protein 1, SDR48A1, short chain dehydrogenase/reductase family 48A, member 1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human NMRAL1. | |
0.1 mL | |
Cancer, Endocrinology, Signal Transduction | |
57407 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction