Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NMRAL1 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306073PE
Description
NMRAL1 Polyclonal antibody specifically detects NMRAL1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| NMRAL1 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| HSCARGFLJ25918, NmrA-like family domain containing 1, nmrA-like family domain-containing protein 1, SDR48A1, short chain dehydrogenase/reductase family 48A, member 1 | |
| Produced in rabbits immunized with E. coli-derived Human NMRAL1 fragment. | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 57407 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction