Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NMS Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101714
Description
Antibody detects endogenous levels of total NMS.
Implicated in the regulation of circadian rhythms through autocrine and/or paracrine actions. {ECO:0000250}.
Specifications
NMS | |
Polyclonal | |
Unconjugated | |
NMS | |
neuromedin S; neuromedin-S; NMS; prepro-NMS | |
Rabbit | |
Affinity chromatography | |
RUO | |
129521 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q5H8A3 | |
NMS | |
A synthesized peptide derived from human NMS(Accession Q5H8A3), corresponding to amino acid residues A118-W153. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction