Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NMT1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | NMT1 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
NMT1 Polyclonal specifically detects NMT1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
NMT1 | |
Polyclonal | |
Rabbit | |
Cancer | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
4836 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:MADESETAVKPPAPPLPQMMEGNGNGHEHCSDCENEEDNSYNRGGLSPANDTGAK | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
alternative, short form NMT-S, glycylpeptide N-tetradecanoyltransferase 1, long form, NMT-L, Myristoyl-CoA:protein N-myristoyltransferase 1, NMT 1, NMTEC 2.3.1.97, N-myristoyltransferase 1, Peptide N-myristoyltransferase 1, Type I N-myristoyltransferase | |
NMT1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title