Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NMUR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113461
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113461 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113461 Supplier Invitrogen™ Supplier No. PA5113461
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NMUR1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Receptor for the neuromedin-U and neuromedin-S neuropeptides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMUR1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene NMUR1
Gene Accession No. O55040, Q9HB89
Gene Alias (FM-3); FM3; FM-3; G protein-coupled receptor 66; G protein-coupled receptor FM-3; GPC-R; GPR66; G-protein coupled receptor 66; G-protein coupled receptor FM-3; neuromedin U receptor 1; neuromedin-U receptor 1; NMU1R; NmU-R; NMUR1; NMU-R1
Gene Symbols NMUR1
Host Species Rabbit
Immunogen Recombinant Human Neuromedin-U receptor 1 protein (360-426AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10316, 14767
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.