Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NMUR1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA530376
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530376 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530376 Supplier Thermo Scientific Supplier No. PA530376

Rabbit Polyclonal Antibody

Recommended positive controls: PC-3, rat testis. Predicted reactivity: Human (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Receptor for the neuromedin-U and neuromedin-S neuropeptides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMUR1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene NMUR1
Gene Accession No. Q9HB89, Q9JJI5
Gene Alias (FM-3); FM3; FM-3; G protein-coupled receptor 66; G protein-coupled receptor FM-3; GPC-R; GPR66; G-protein coupled receptor 66; G-protein coupled receptor FM-3; neuromedin U receptor 1; neuromedin-U receptor 1; NMU1R; NmU-R; NMUR1; NMU-R1
Gene Symbols NMUR1
Host Species Rabbit
Immunogen Recombinant protein fragment corresponding to a region within amino acids 88 and 376 of Human NMUR1
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10316, 65276
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.