Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NMUR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588771
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588771 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588771 Supplier Invitrogen™ Supplier No. PA588771
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IYNLLSRRFQ AAFQNVISSF HKQWHSQHDP QLPPAQRNIF LTECHFVELT EDIGPQFPCQ SSMHNSHLPA ALSSEQMSRT NYQSFHFNKT; Positive Samples: SGC-7901, Mouse uterus, Mouse small intestine, Rat uterus; Cellular Location: Cell membrane, Multi-pass membrane protein.

This gene encodes a protein from the G-protein coupled receptor 1 family. This protein is a receptor for neuromedin U, which is a neuropeptide that is widely distributed in the gut and central nervous system. This receptor plays an important role in the regulation of food intake and body weight.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMUR2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Nmur2
Gene Accession No. Q8BZ39, Q9ESQ4, Q9GZQ4
Gene Alias FM4; FM-4; G-protein coupled receptor FM-4; G-protein coupled receptor TGR-1; G-protein-coupled receptor FM-4; growth hormone secretagogue receptor family, member 4; neuromedin s receptor 2 protein; neuromedin U receptor 2; neuromedin-U receptor 2; NMSR2; NMU2R; NMUR2; NMU-R2; Tgr1; TGR-1
Gene Symbols Nmur2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 326-415 of human NMUR2 (NP_0645523).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216749, 56923, 64042
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.