Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NMUR2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568683
Description
This target displays homology in the following species: Human: 100%.
This gene encodes a protein from the G-protein coupled receptor 1 family. This protein is a receptor for neuromedin U, which is a neuropeptide that is widely distributed in the gut and central nervous system. This receptor plays an important role in the regulation of food intake and body weight.
Specifications
NMUR2 | |
Polyclonal | |
Unconjugated | |
Nmur2 | |
FM4; FM-4; G-protein coupled receptor FM-4; G-protein coupled receptor TGR-1; G-protein-coupled receptor FM-4; growth hormone secretagogue receptor family, member 4; neuromedin s receptor 2 protein; neuromedin U receptor 2; neuromedin-U receptor 2; NMSR2; NMU2R; NMUR2; NMU-R2; Tgr1; TGR-1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
56923 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9GZQ4 | |
Nmur2 | |
synthetic peptide directed towards the N terminal region of human NMUR2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction