Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NNC 0640
GREENER_CHOICE

Catalog No. 680710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-071-0 10 mg
68-075-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 68-071-0 Supplier Tocris Bioscience™ Supplier No. 6807/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Negative allosteric modulator of glucagon and GLP-1 receptors

NNC 0640 is a negative allosteric modulator of glucagon receptors (pKi = 7.4). Binds to external surface of the transmembrane domain. Also negative allosteric modulator of glucagon-like peptide 1 (GLP-1) receptor. Inhibits GLP-1-mediated cAMP accumulation in vitro.

Specifications

Chemical Name or Material 4-[[(4-Cyclohexylphenyl)[[[3-(methylsulfonyl)phenyl]amino]carbonyl]amino]methyl]-N-2 H-tetrazol-5-ylbenzamide
CAS 307986-98-7
Quantity 10 mg
Target Glucagon-Like Peptide 1 Receptor Modulators
Molecular Formula C29H31N7O4S
Formula Weight 2172
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.