Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NNF1R Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988984
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8984 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8984 Supplier Bioss Supplier No. BS9319R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PMF1 is part of the MIS12 complex which is required for normal chromosome alignment and segregation and kinetochore formation during mitosis. It may act as a cotranscription partner of NFE2L2 involved in regulation of polyamine-induced transcription of SSAT.

Specifications

Antigen NNF1R
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PMF1
Gene Accession No. Q6P1K2
Gene Alias 2600009M07Rik; AL033286; AW060657; BOS_2831; Est1p-like protein B (EST1B); LOC100341536; PMF1; PMF-1; polyamine modulated factor 1; Polyamine-modulated factor 1; polyamine-modulated factor 1-like; RP11-54H19.4
Gene Symbols PMF1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NNF1R/PMF1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11243
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.