Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NNMT Monoclonal Antibody (OTI1A1), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502470

Description
N-methylation is one method by which drug and other xenobiotic compounds are metabolized by the liver. This gene encodes the protein responsible for this enzymatic activity which uses S-adenosyl methionine as the methyl donor.Specifications
NNMT | |
Monoclonal | |
Unconjugated | |
NNMT | |
nicotinamide N-methyltransferase | |
Mouse | |
Affinity Chromatography | |
RUO | |
4837, 489396 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1A1 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P40261 | |
NNMT | |
Full length human recombit protein of human NNMT produced in HEK293T cell. | |
100 μL | |
Primary | |
Canine, Human | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction