Learn More
NNMT Mouse anti-Human, Clone: OTI2G8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502522

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
N-methylation is one method by which drug and other xenobiotic compounds are metabolized by the liver. This gene encodes the protein responsible for this enzymatic activity which uses S-adenosyl methionine as the methyl donor.Specifications
NNMT | |
Monoclonal | |
Unconjugated | |
NNMT | |
nicotinamide N-methyltransferase | |
Mouse | |
Affinity Chromatography | |
RUO | |
4837 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2G8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P40261 | |
NNMT | |
Full length human recombit protein of human NNMT produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.