Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NNMT Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598011
Description
NNMT Polyclonal Antibody for Western Blot, IHC (P), ELISA
N-methylation is one method by which drug and other xenobiotic compounds are metabolized by the liver. This gene encodes the protein responsible for this enzymatic activity which uses S-adenosyl methionine as the methyl donor.
Specifications
NNMT | |
Polyclonal | |
Unconjugated | |
NNMT | |
Nicotinamide N-methyltransferase; NNMT | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4837 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P40261 | |
NNMT | |
Recombinant Human Nicotinamide N-methyltransferase protein (1-264AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction