Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NNMT Polyclonal Antibody

Catalog No. PIPA5121872
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121872 20 μL
PIPA5119308 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121872 Supplier Invitrogen™ Supplier No. PA5121872
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NNMT Polyclonal Antibody for Western Blot, IHC (P)

N-methylation is one method by which drug and other xenobiotic compounds are metabolized by the liver. This gene encodes the protein responsible for this enzymatic activity which uses S-adenosyl methionine as the methyl donor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NNMT
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene NNMT
Gene Accession No. P40261
Gene Alias Nicotinamide N-methyltransferase; NNMT
Gene Symbols NNMT
Host Species Rabbit
Immunogen Recombinant protein Nicotinamide-N-Methyltransferase. The antigen corresponds to amino acid range 1-258 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4837
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.