Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
nNOS Antibody (60718), Alexa Fluor 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals MAB2416AF532
Description
nNOS Monoclonal specifically detects nNOS in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
nNOS | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
NOS1 | |
Partial recombinant human nNOS protein (amino acids 218-1434) [UniProt P29475] | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
60718 | |
Western Blot | |
bNOS, brain, Constitutive NOS, EC 1.14.13.39, IHPS1, Neuronal NOS, nitric oxide synthase 1 (neuronal), N-NOS, nNOSNOS type I, NOS, Peptidyl-cysteine S-nitrosylase NOS1 | |
Mouse | |
0.1 mL | |
4842 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction