Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nNOS Antibody, Novus Biologicals™
SDP

Catalog No. NB100858 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100858 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100858 Supplier Novus Biologicals Supplier No. NB100858
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 12 publications

nNOS Polyclonal specifically detects nNOS in Human, Mouse, Rat, Guinea Pig samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Peptide ELISA, Immunohistochemistry Whole-Mount.

Specifications

Antigen nNOS
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3-1 ug/ml, Flow Cytometry 10 ug/mL, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 10 ug/mL, Immunohistochemistry-Frozen 1:10-1:500, Peptide ELISA detection limit 1:64000, Immunohistochemistry Whole-Mount
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. P29475
Gene Alias bNOS, brain, Constitutive NOS, EC 1.14.13.39, IHPS1, Neuronal NOS, nitric oxide synthase 1 (neuronal), N-NOS, nNOSNOS type I, NOS, Peptidyl-cysteine S-nitrosylase NOS1
Gene Symbols NOS1
Host Species Goat
Immunogen Synthetic peptide with sequence ESKKDTDEVFSS, representing the C-terminus of the human protein (residues 1423-1434) according to NP_000611.
Molecular Weight of Antigen 161 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Asthma, Cancer, HIF Target Genes, Hypoxia, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4842
Test Specificity Expected from sequence similarity: Rat, Dog and Cow.
Target Species Human, Mouse, Rat, Guinea Pig
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.