Learn More
Invitrogen™ nNOS Recombinant Rabbit Monoclonal Antibody (ST520)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA532172
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
NOS activity requires homodimerization as well as three cosubstrates (L-arginine, NADPH and O2) and five cofactors or prosthetic groups (FAD, FMN, calmodulin, tetrahydrobiopterin and heme). Several distinct NOS isoforms have been described and been shown to represent the products of three distinct genes. These include two constitutive Ca2+/CaM-dependent forms of NOS, including NOS1 (also designated ncNOS) whose activity was first identified in neurons, and NOS3 (also designated ecNOS), first identified in endothelial cells.
Specifications
nNOS | |
Recombinant Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
P29475, P29476, Q9Z0J4 | |
NOS1 | |
Synthetic peptide within Human nNOS aa 1398-1434. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
ST520 | |
Unconjugated | |
NOS1 | |
2310005C01Rik; bNOS; bNOS1; Brain NOS; Constitutive NOS; IHPS1; NC-NOS; neuronal nitric oxide synthase; neuronal nitric oxide synthase NOS1; neuronal NOS; nitric oxidase synthase; nitric oxide synthase; nitric oxide synthase 1; nitric oxide synthase 1 (neuronal); nitric oxide synthase 1, neuronal; nitric oxide synthase, brain; nNOS; N-NOS; nNOS; NOS type I; NO; NOS; NOS Type 1; NOS type I; Nos1; Nos-1; NOS-I; Peptidyl-cysteine S-nitrosylase NOS1; Phospho-bNOS; Phospho-Brain NOS | |
Rabbit | |
Protein A | |
RUO | |
18125, 24598, 4842 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.