Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NNP1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988969
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8969 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8969 Supplier Bioss Supplier No. BS9300R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is the putative homolog of the yeast ribosomal RNA processing protein RRP1. The encoded protein is involved in the late stages of nucleologenesis at the end of mitosis, and may be required for the generation of 28S rRNA.

Specifications

Antigen NNP1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RRP1
Gene Accession No. P56182
Gene Alias D21S2056E; NNP1; NNP-1; Nnp1 homolog, nucleolar protein; NOP52; Novel nuclear protein 1; Nucleolar protein Nop52; protein NNP-1; ribosomal RNA processing 1; ribosomal RNA processing 1 homolog; Ribosomal RNA processing protein 1 homolog A; RRP1; RRP1A; RRP1-like protein
Gene Symbols RRP1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NNP1/RRP1 like protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8568
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.