Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NNT Monoclonal Antibody (8B4BB10)
GREENER_CHOICE

Catalog No. 459170
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-917-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-917-0 Supplier Invitrogen™ Supplier No. 459170
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Normal Human skin fibroblasts (strain HDFn); Human cerebellum tissue; mitochondria from Human liver, Bovine heart, Rat liver and Mouse liver; HepG2 and HL-60 cells.

This gene encodes an integral protein of the inner mitochondrial membrane. The enzyme couples hydride transfer between NAD(H) and NADP(+) to proton translocation across the inner mitochondrial membrane. Under most physiological conditions, the enzyme uses energy from the mitochondrial proton gradient to produce high concentrations of NADPH. The resulting NADPH is used for biosynthesis and in free radical detoxification.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NNT
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 8B4BB10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation HEPES buffered saline with 0.02% sodium azide
Gene NNT
Gene Accession No. P11024, Q13423, Q61941
Gene Alias 4930423F13Rik; AI323702; BB168308; GCCD4; NAD(P) transhydrogenase, mitochondrial; nicotinamide nucleotide transhydrogenase; Nicotinamide nucleotide transhydrogenase (NAD(P)+ transhydrogenase); NNT; Pyridine nucleotide transhydrogenase; pyridine nucleotide transhydrogenase precursor (EC 1.6.1.1)
Gene Symbols NNT
Host Species Mouse
Immunogen Human heart mitochondria.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18115, 23530, 280878, 310378
Target Species Human, Mouse, Rat, Bovine
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.