Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NNT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596293
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596293 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596293 Supplier Invitrogen™ Supplier No. PA596293
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NRSLANVILG GYGTTSTAGG KPMEISGTHT EINLDNAIDM IREANSIIIT PGYGLCAAKA QYPIADLVKM LTEQGKKVRF GIHPVAGRMP GQLNVLLAEA GVPYDIVLEM DEINHDFPDT DLVLVIGAND TVNSAAQEDP NSIIAGMPVL EVWKSKQVIV MKRSLGVGYA AVDNPIFYKP NTAMLLGDAK KTCDALQAKV RESYQK; Positive Samples: HepG2, Mouse heart, Mouse liver, Rat kidney; Cellular Location: Matrix side, Mitochondrion inner membrane, Multi-pass membrane protein.

This gene encodes an integral protein of the inner mitochondrial membrane. The enzyme couples hydride transfer between NAD(H) and NADP(+) to proton translocation across the inner mitochondrial membrane. Under most physiological conditions, the enzyme uses energy from the mitochondrial proton gradient to produce high concentrations of NADPH. The resulting NADPH is used for biosynthesis and in free radical detoxification.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NNT
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.88 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NNT
Gene Accession No. Q13423, Q61941
Gene Alias 4930423F13Rik; AI323702; BB168308; GCCD4; NAD(P) transhydrogenase, mitochondrial; nicotinamide nucleotide transhydrogenase; Nicotinamide nucleotide transhydrogenase (NAD(P)+ transhydrogenase); NNT; Pyridine nucleotide transhydrogenase; pyridine nucleotide transhydrogenase precursor (EC 1.6.1.1)
Gene Symbols NNT
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 881-1086 of human NNT (NP_0364753).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18115, 23530, 310378
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.