Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NO66/C14orf169 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 228421AP150UL
Description
Histone demethylase that specifically demethylates ′Lys-4′ (H3K4me) and ′Lys-36′ (H3K36me) of histone H3, thereby playing a central role in histone code. Preferentially demethylates trimethylated H3 ′Lys-4′ (H3K4me3) and monomethylated H3 ′Lys-4′ (H3K4me1) residues, while it has weaker activity for dimethylated H3 ′Lys-36′ (H3K36me2). Acts as a regulator of osteoblast differentiation via its interaction with SP7/OSX by demethylating H3K4me and H3K36me, thereby inhibiting SP7/OSX-mediated promoter activation. May also play a role in ribosome biogenesis and in the replication or remodeling of certain heterochromatic region.Specifications
| NO66/C14orf169 | |
| Polyclonal | |
| Unconjugated | |
| RIOX1 | |
| C14orf169, hsNO66, MAPJD, NO66, Nucleolar protein 66 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 79697 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H6W3 | |
| RIOX1 | |
| NO66/C14orf169 Fusion Protein Ag18721 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction