Learn More
NO66 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303198AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Histone demethylase that specifically demethylates ′Lys-4′ (H3K4me) and ′Lys-36′ (H3K36me) of histone H3, thereby playing a central role in histone code. Preferentially demethylates trimethylated H3 ′Lys-4′ (H3K4me3) and monomethylated H3 ′Lys-4′ (H3K4me1) residues, while it has weaker activity for dimethylated H3 ′Lys-36′ (H3K36me2). Acts as a regulator of osteoblast differentiation via its interaction with SP7/OSX by demethylating H3K4me and H3K36me, thereby inhibiting SP7/OSX-mediated promoter activation. May also play a role in ribosome biogenesis and in the replication or remodeling of certain heterochromatic region.Specifications
NO66 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9H6W3 | |
RIOX1 | |
Between 1 and 50 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
RIOX1 | |
C14orf169; NO66; chromosome 14 open reading frame 169; FLJ21802; MAPJD | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79697 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.