Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOA1 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299264MFV450
Description
NOA1 Polyclonal antibody specifically detects NOA1 in Human samples. It is validated for Western BlotSpecifications
NOA1 | |
Polyclonal | |
Western Blot | |
chromosome 4 open reading frame 14, hAtNOS1, mAtNOS1, MGC3232, mitochondrial nitric oxide synthase, nitric oxide synthase, mitochondrial (putative), putative ortholog of Arabidopsis mitochondrial nitric oxide synthase | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human NOA1. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
84273 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction