Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOB1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | NOB1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
NOB1 Polyclonal specifically detects NOB1 in Human samples. It is validated for Western Blot.Specifications
NOB1 | |
Polyclonal | |
Rabbit | |
Q9ULX3 | |
28987 | |
Synthetic peptides corresponding to NOB1(NIN1/RPN12 binding protein 1 homolog (S. cerevisiae)) The peptide sequence was selected from the N terminal of NOB1. Peptide sequence TEIRDKATRRRLAVLPYELRFKEPLPEYVRLVTEFSKKTGDYPSLSATDI. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
adenocarcinoma antigen recognized by T lymphocytes 4, ART4, ART-4, MST158, nin one binding protein, NIN1/RPN12 binding protein 1 homolog (S. cerevisiae), NOB1PPSMD8BP1, Phosphorylation regulatory protein HP-10, Protein ART-4, PSMD8 binding protein 1, RNA-binding protein NOB1 | |
NOB1 | |
IgG | |
47 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title