Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOB1 Antibody (OTI1C12), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP246187
Description
Ensure accurate, reproducible results in Western Blot
NOB1 Monoclonal specifically detects NOB1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
NOB1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000 | |
adenocarcinoma antigen recognized by T lymphocytes 4, ART4, ART-4, MST158, nin one binding protein, NIN1/RPN12 binding protein 1 homolog (S. cerevisiae), NOB1PPSMD8BP1, Phosphorylation regulatory protein HP-10, Protein ART-4, PSMD8 binding protein 1, RNA-binding protein NOB1 | |
Mouse | |
46.5 kDa | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
28987 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot | |
OTI1C12 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
NOB1 | |
Human recombinant protein fragment corresponding to amino acids 2-234 of human NOB1(NP_054781) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction