Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NOB1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561967
Description
Immunogen sequence: PSNIKQIQQE LEQCDVPEDV RVGCLTTDFA MQNVLLQMGL HVLAVNGMLI REARSYILRC HGCFKTTSDM SRVFCSHCGN KTLKKVSVTV SDDGTLHMHF SR Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 89%.
In yeast, over 200 protein and RNA cofactors are required for ribosome assembly, and these are generally conserved in eukaryotes. These factors orchestrate modification and cleavage of the initial 35S precursor rRNA transcript into the mature 18S, 5.8S, and 25S rRNAs, folding of the rRNA, and binding of ribosomal proteins and 5S RNA. Nob1 is involved in pre-rRNA processing. In a late cytoplasmic processing step, Nob1 cleaves a 20S rRNA intermediate at cleavage site D to produce the mature 18S rRNA.
Specifications
NOB1 | |
Polyclonal | |
Unconjugated | |
Nob1 | |
1700021I09Rik; 20S-pre-rRNA D-site endonuclease NOB1; adenocarcinoma antigen recognized by T lymphocytes 4; ART4; ART-4; MST158; MSTP158; nin one binding protein; NIN1/PSMD8 binding protein 1 homolog; NIN1/RPN12 binding protein 1 homolog; NIN1/RPN12 binding protein 1 homolog (S. cerevisiae); NIN1-binding protein; Nob1; Nob1p; phosphorylation regulatory protein HP-10; Pre-rRNA-processing endonuclease NOB1; Protein ART-4; PSMD8 binding protein 1; PSMD8BP1; RNA-binding protein NOB1; rRNA-binding endoribonuclease; YOR056C; YOR29-07 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
28987 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9ULX3 | |
Nob1 | |
Recombinant protein corresponding to Human NOB1. Recombinant protein control fragment (Product #RP-98724). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction