Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110036
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110036 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110036 Supplier Invitrogen™ Supplier No. PA5110036
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAPVEHVVAD AGAFLRHAAL QDIGKNIYTI REVVTEIRDK ATRRRLAVLP YELRFKEPLP EYVRLVTEFS KKTGDYPSLS ATDIQVLALT YQLEAEFVGV SHLKQEPQKV KVSSSIQHPE TPLHISGFHL PYKPKPPQET EKGHSACEPE NLEFSSFMFW RNPLPNIDHE LQELLIDRGE DVPSEEEEEE ENGFEDRKDD SDDDGGGWIT PSNIKQIQQE LEQCDVPEDV.

In yeast, over 200 protein and RNA cofactors are required for ribosome assembly, and these are generally conserved in eukaryotes. These factors orchestrate modification and cleavage of the initial 35S precursor rRNA transcript into the mature 18S, 5.8S, and 25S rRNAs, folding of the rRNA, and binding of ribosomal proteins and 5S RNA. Nob1 is involved in pre-rRNA processing. In a late cytoplasmic processing step, Nob1 cleaves a 20S rRNA intermediate at cleavage site D to produce the mature 18S rRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOB1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.17 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Nob1
Gene Accession No. Q6VEU1, Q8BW10, Q9ULX3
Gene Alias 1700021I09Rik; 20S-pre-rRNA D-site endonuclease NOB1; adenocarcinoma antigen recognized by T lymphocytes 4; ART4; ART-4; MST158; MSTP158; nin one binding protein; NIN1/PSMD8 binding protein 1 homolog; NIN1/RPN12 binding protein 1 homolog; NIN1/RPN12 binding protein 1 homolog (S. cerevisiae); NIN1-binding protein; Nob1; Nob1p; phosphorylation regulatory protein HP-10; Pre-rRNA-processing endonuclease NOB1; Protein ART-4; PSMD8 binding protein 1; PSMD8BP1; RNA-binding protein NOB1; rRNA-binding endoribonuclease; YOR056C; YOR29-07
Gene Symbols Nob1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human NOB1 (NP_054781.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28987, 291996, 67619
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.