Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOBOX Antibody (OTI4C5), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP273028UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273028UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273028UV Supplier Novus Biologicals Supplier No. NBP273028UV
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NOBOX Monoclonal specifically detects NOBOX in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen NOBOX
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4C5
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias NOBOX NOBOX oogenesis homeobox, OG2, OG-2, OG2X, POF5, TCAG_12042
Gene Symbols NOBOX
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-271 of human NOBOX (NP_001073882) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 135935.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.